Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmigd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025655.2 | NP_079931.1 | 3, 4 | 658 | ||
GenBank mRNA | AK008028.1 | - | 4, 5 | 719 | ||
AK009118.1 | - | 3, 4 | 658 | |||
BC022683.1 | - | 3, 4 | 625 | |||
BC061082.1 | - | 3, 4 | 648 | 30311543 |