Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025656.4 | NP_079932.2 | 7, 8 | 693 | ||
GenBank mRNA | AK005928.1 | - | 7, 8 | 695 | ||
AK007515.1 | - | 7, 8 | 694 | |||
AK013414.1 | - | 7, 8 | 650 | |||
BC053424.1 | - | 7, 8 | 1122 | 6336522 |