Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174874.3 | NP_777363.1 | 5 | 386 | ||
GenBank mRNA | AJ504653.1 | - | 4 | 378 | ||
AK010949.1 | - | 5 | 514 | |||
AK028712.1 | - | 5 | 397 | |||
AK031570.1 | - | 5 | 387 | |||
AK075796.1 | - | 5 | 397 | |||
AK088811.1 | - | 5 | 459 | |||
AK129242.1 | - | 4 | 374 | |||
AK144239.1 | - | 5 | 392 | |||
AK188147.1 | - | 3 | 235 | |||
AK204065.1 | - | 1 | 74 | |||
BC027184.1 | - | 5 | 412 | 4237950 |