Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025664.5 | NP_079940.2 | 7 | 832 | ||
GenBank mRNA | AK012520.1 | - | 2 | 124 | ||
AK139802.1 | - | 7 | 866 | |||
AK149739.1 | - | 7 | 846 | |||
AK149849.1 | - | 7 | 886 | |||
AK155830.1 | - | 867 | ||||
AK157517.1 | - | 7 | 848 | |||
AK170339.1 | - | 848 | ||||
AK191492.1 | - | 3 | 363 | |||
BC014814.1 | - | 7 | 832 | 3257422 |