Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025664.5 | NP_079940.2 | 8, 9 | 989 | ||
GenBank mRNA | AK012520.1 | - | 3, 4 | 281 | ||
AK139802.1 | - | 8, 9 | 1023 | |||
AK149739.1 | - | 8, 9 | 1003 | |||
AK149849.1 | - | 8, 9 | 1043 | |||
AK155830.1 | - | 1024 | ||||
AK157517.1 | - | 8, 9 | 1005 | |||
AK170339.1 | - | 1005 | ||||
BC014814.1 | - | 8, 9 | 989 | 3257422 |