Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114937.2 | NP_001108409.1 | 2 | 532 | ||
NM_002351.4 | NP_002342.1 | 2 | 532 | |||
GenBank mRNA | AB586694.1 | - | 2 | 273 | ||
AF072930.1 | - | 2 | 257 | |||
AF073019.1 | - | 2 | 257 | |||
AF100539.1 | - | 2 | 225 | |||
AF100541.1 | - | 2 | 225 | |||
AF100542.1 | - | 2 | 203 | |||
AK311911.1 | - | 2 | 206 | |||
AL023657.1 | - | 2 | 470 | |||
BC020732.1 | - | 2 | 264 | 4702653 | ||
CR542031.1 | - | 2 | 171 | |||
CR542043.1 | - | 2 | 171 | |||
DA007747.1 | - | 2 | 532 | |||
JX512648.1 | - | 2 | 204 |