Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025673.2 | NP_079949.1 | 2 | 515 | ||
GenBank mRNA | AK005996.1 | - | 2 | 562 | ||
AK014644.1 | - | 2 | 563 | |||
AK015543.1 | - | 2 | 559 | |||
AK021327.1 | - | 2 | 561 | |||
AK028409.1 | - | 2 | 524 | |||
AK077446.1 | - | 2 | 580 | |||
AK133763.1 | - | 2 | 563 | |||
AK136194.1 | - | 2 | 140 | |||
AK139545.1 | - | 2 | 562 | |||
AK167805.1 | - | 2 | 556 | |||
BC031445.1 | - | 2 | 376 | 4482612 | ||
BC039909.1 | - | 2 | 513 | |||
BC051035.1 | - | 2 | 515 | 2655316 |