Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025675.4 | NP_079951.1 | 4 | 443 | ||
GenBank mRNA | AK006965.1 | - | 4 | 401 | ||
AK007021.1 | - | 4 | 446 | |||
AK017595.1 | - | 4 | 399 | |||
AK036021.1 | - | 4 | 479 | |||
AK082013.1 | - | 4 | 480 | |||
AK088160.1 | - | 4 | 445 | |||
AK145789.1 | - | 4 | 403 | |||
BC022995.1 | - | 4 | 468 | 5363234 |