Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000239.2 | NP_000230.1 | 3 | 404 | ||
GenBank mRNA | AK130149.1 | - | 3 | 409 | ||
AK310930.1 | - | 286 | ||||
AK311779.1 | - | 3 | 377 | |||
BC004147.2 | - | 3 | 368 | 2959387 | ||
FJ795023.1 | - | 3 | 371 | |||
J03801.1 | - | 3 | 374 | |||
KJ841928.1 | - | 3 | 349 | |||
M19045.1 | - | 3 | 362 | |||
M21119.1 | - | 3 | 362 | |||
U25677.1 | - | 3 | 349 |