Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025681.2 | NP_079957.2 | 4 | 704 | ||
GenBank mRNA | AF351204.1 | - | 4 | 445 | ||
AK012220.1 | - | 4 | 512 | |||
AK017685.1 | - | 4 | 523 | |||
BC049574.1 | - | 4 | 718 | 6593108 | ||
BC052700.1 | - | 4 | 504 | 6416835 | ||
BC063057.1 | - | 4 | 497 | 5715784 |