Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025681.2 | NP_079957.2 | 6 | 849 | ||
GenBank mRNA | AK012220.1 | - | 6 | 657 | ||
AK017685.1 | - | 6 | 668 | |||
AK161930.1 | - | 3 | 426 | |||
BC049574.1 | - | 6 | 863 | 6593108 | ||
BC052700.1 | - | 6 | 649 | 6416835 | ||
BC063057.1 | - | 6 | 642 | 5715784 |