Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019800.4 | NP_062774.2 | 9 | 1448 | ||
GenBank mRNA | AB030038.1 | - | 9 | 1441 | ||
AF216223.1 | - | 9 | 925 | |||
AK002783.1 | - | 9 | 1362 | |||
AK045011.1 | - | 1721 | ||||
AK077729.1 | - | 9 | 1436 | |||
AK160271.1 | - | 9 | 1367 | |||
AK165356.1 | - | 9 | 1449 | |||
AK171757.1 | - | 9 | 1363 | |||
BC023190.1 | - | 2 | 1351 | |||
BC047276.1 | - | 9 | 1188 | 6487551 |