Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025690.3 | NP_079966.2 | 15 | 2113 | ||
NM_026337.1 | NP_080613.1 | 15 | 2151 | |||
GenBank mRNA | AF462146.1 | - | 15 | 2113 | ||
AK045723.1 | - | 9 | 1213 | |||
AK083079.1 | - | 1 | 2641 | |||
AK134756.1 | - | 6 | 792 | |||
BC019992.1 | - | 4 | 483 | |||
BC139202.1 | - | 15 | 2103 | 8862224 | ||
CD806976.1 | - | 3 | 354 |