Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025690.3 | NP_079966.2 | 1, 2 | 210 | ||
NM_026337.1 | NP_080613.1 | 1, 2 | 302 | |||
GenBank mRNA | AF462146.1 | - | 1, 2 | 210 | ||
AK011815.1 | - | 1, 2 | 265 | |||
AK017839.1 | - | 1, 2 | 163 | |||
AK077578.1 | - | 1, 2 | 325 | |||
AK078251.1 | - | 1, 2 | 287 | |||
AK162780.1 | - | 1, 2 | 183 | |||
BC139202.1 | - | 1, 2 | 200 | 8862224 | ||
BC145577.1 | - | 1, 2 | 200 |