Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025691.1 | NP_079967.1 | 13 | 1324 | ||
GenBank mRNA | AK133784.1 | - | 13 | 1325 | ||
AK164641.1 | - | 7 | 682 | |||
BC003434.1 | - | 13 | 1312 | |||
BC019196.1 | - | 13 | 1317 | |||
BC089462.1 | - | 4 | 334 | |||
BY393170.1 | - | 1 | 45 |