Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025692.3 | NP_079968.2 | 2 | 359 | ||
GenBank mRNA | AK017787.1 | - | 2 | 359 | ||
AK077825.1 | - | 2 | 341 | |||
AK151823.1 | - | 2 | 316 | |||
AK152029.1 | - | 2 | 354 | |||
AK168907.1 | - | 2 | 353 | |||
AK206146.1 | - | 2 | 76 | |||
BC019764.1 | - | 2 | 236 | 3989119 |