Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175112.5 | NP_780321.1 | 5 | 580 | ||
GenBank mRNA | AK014325.1 | - | 5 | 537 | ||
AK017746.1 | - | 5 | 501 | |||
AK035278.1 | - | 5 | 526 | |||
AK079383.1 | - | 5 | 561 | |||
AK146058.1 | - | 5 | 545 | |||
AK156681.1 | - | 5 | 545 | |||
AK209630.1 | - | 2 | 43 | |||
AK215054.1 | - | 2 | 43 | |||
BC059051.1 | - | 5 | 502 | 6818771 | ||
BC060072.1 | - | 5 | 447 | 5718934 |