Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025699.2 | NP_079975.2 | 4 | 800 | ||
GenBank mRNA | AK006969.1 | - | 4 | 843 | ||
AK009708.1 | - | 4 | 816 | |||
AK014323.1 | - | 4 | 819 | |||
AK088622.1 | - | 4 | 819 | |||
AK139692.1 | - | 972 | ||||
AK153499.1 | - | 4 | 837 | |||
BC005579.1 | - | 4 | 800 | 3968876 |