Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025701.4 | NP_079977.3 | 2 | 647 | ||
GenBank mRNA | AK003633.1 | - | 2 | 615 | ||
AK007989.1 | - | 2 | 614 | |||
AK076212.1 | - | 2 | 648 | |||
AK078451.1 | - | 2 | 613 | |||
AK171833.1 | - | 1 | 3027 | |||
BC028946.1 | - | 1 | 1107 | |||
BC049179.1 | - | 2 | 605 | 6313540 |