Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025706.3 | NP_079982.3 | 11 | 1334 | ||
GenBank mRNA | AK014477.1 | - | 11 | 1266 | ||
AK014795.1 | - | 6 | 717 | |||
AK078875.1 | - | 11 | 1280 | |||
AK147085.1 | - | 11 | 1297 | |||
BC032087.1 | - | 11 | 1262 | |||
BC053395.1 | - | 11 | 1315 | 30054233 | ||
DQ054831.1 | - | 11 | 1265 |