Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapvd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025709.2 | NP_079985.2 | 11 | 2419 | ||
GenBank mRNA | AK129381.1 | - | 10 | 2014 | ||
AK134776.1 | - | 9 | 1409 | |||
AK150026.1 | - | 10 | 2028 | |||
BC031478.1 | - | 8 | 1464 | |||
BC032088.1 | - | 8 | 1464 | |||
BC048847.1 | - | 10 | 1985 | |||
BC060123.1 | - | 11 | 2419 | 6408778 | ||
EF155419.1 | - | 9 | 1837 |