Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapvd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025709.2 | NP_079985.2 | 3 | 718 | ||
GenBank mRNA | AK129381.1 | - | 2 | 313 | ||
AK150026.1 | - | 2 | 327 | |||
AK165047.1 | - | 2 | 330 | |||
BC048847.1 | - | 2 | 284 | |||
BC060123.1 | - | 3 | 718 | 6408778 | ||
EF155419.1 | - | 1 | 136 |