Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025712.4 | NP_079988.3 | 10 | 963 | ||
GenBank mRNA | AK014536.1 | - | 10 | 965 | ||
AK034196.1 | - | 10 | 965 | |||
AK035226.1 | - | 10 | 964 | |||
BC061007.1 | - | 10 | 959 | 30314775 |