Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Henmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078646.1 | NP_001072114.1 | 5 | 529 | ||
NM_025723.2 | NP_079999.2 | 5 | 844 | |||
GenBank mRNA | AK014900.1 | - | 5 | 810 | ||
AK038994.1 | - | 5 | 530 | |||
AK085587.1 | - | 5 | 844 | |||
BC099408.1 | - | 4 | 510 | 30935249 | ||
BC116412.1 | - | 3 | 325 | |||
BC116413.1 | - | 3 | 325 | |||
BC128494.1 | - | 3 | 325 | |||
BC128495.1 | - | 3 | 325 |