Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025727.3 | NP_080003.1 | 5 | 1889 | ||
GenBank mRNA | AK006288.1 | - | 5 | 1876 | ||
AK014906.1 | - | 5 | 1890 | |||
AK016593.1 | - | 3 | 955 | |||
AK077202.1 | - | 5 | 1891 | |||
AY495337.1 | - | 5 | 1890 | |||
BC139267.1 | - | 5 | 1865 | 8862289 | ||
BC139268.1 | - | 5 | 1865 | 8862290 |