Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025728.4 | NP_080004.1 | 2 | 246 | ||
NM_029160.3 | NP_083436.2 | 11 | 1087 | |||
GenBank mRNA | AF490391.1 | - | 11 | 1069 | ||
AK014865.1 | - | 2 | 241 | |||
AK015884.1 | - | 11 | 1088 | |||
AK016352.1 | - | 2 | 214 | |||
AK016618.1 | - | 2 | 233 | |||
BC120668.1 | - | 11 | 1086 | 40129591 | ||
BC145006.1 | - | 11 | 1086 | 9053554 | ||
CJ046986.1 | - | 2 | 247 |