Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933415F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025746.2 | NP_080022.2 | 1 | 333 | ||
GenBank mRNA | AK016819.1 | - | 1 | 334 | ||
BC115556.1 | - | 1 | 334 | 40101816 |