Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025747.3 | NP_080023.1 | 1 | 1045 | ||
GenBank mRNA | AK016776.1 | - | 2 | 1063 | ||
AK048125.1 | - | 2 | 1130 | |||
AK082519.1 | - | 3 | 1237 | |||
AK129001.1 | - | 1 | 480 | |||
AK129358.1 | - | 2 | 1103 | |||
BC049135.1 | - | 2 | 1088 | 6831539 | ||
BC058092.1 | - | 1 | 1047 | 6821844 |