Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933425L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025751.3 | NP_080027.1 | 7 | 1506 | ||
GenBank mRNA | AK016761.1 | - | 7 | 1502 | ||
AK016919.1 | - | 7 | 1507 | |||
BC119576.1 | - | 7 | 1494 | 40058060 | ||
BC119577.1 | - | 7 | 1494 | 40058061 |