Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025768.2 | NP_080044.2 | 4 | 485 | ||
GenBank mRNA | AF329833.1 | - | 2 | 273 | ||
AK007905.1 | - | 4 | 490 | |||
AK017197.1 | - | 4 | 449 | |||
AK017251.1 | - | 4 | 396 | |||
AK044437.1 | - | 449 | ||||
AK082863.1 | - | 4 | 479 | |||
AK149322.1 | - | 482 | ||||
BC029096.1 | - | 4 | 491 | 3500563 |