Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025773.3 | NP_080049.2 | 4 | 310 | ||
GenBank mRNA | AK015348.1 | - | 4 | 264 | ||
AK042870.1 | - | 4 | 260 | |||
AK044463.1 | - | 4 | 262 | |||
AK046086.1 | - | 4 | 264 | |||
AK050679.1 | - | 4 | 262 | |||
AK077997.1 | - | 4 | 235 | |||
AK082233.1 | - | 4 | 259 | |||
BC020124.1 | - | 4 | 262 | 4012785 | ||
BY728312.1 | - | 4 | 310 | |||
DV044691.1 | - | 2 | 156 |