Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271016.1 | NP_001257945.1 | 2 | 164 | ||
NM_001271017.1 | NP_001257946.1 | 2 | 164 | |||
NM_025773.3 | NP_080049.2 | 2 | 164 | |||
GenBank mRNA | AK015348.1 | - | 2 | 118 | ||
AK042870.1 | - | 2 | 114 | |||
AK044463.1 | - | 2 | 116 | |||
AK046086.1 | - | 2 | 118 | |||
AK050679.1 | - | 2 | 116 | |||
AK050923.1 | - | 2 | 138 | |||
AK077997.1 | - | 2 | 89 | |||
AK082233.1 | - | 2 | 113 | |||
BC020124.1 | - | 2 | 116 | 4012785 | ||
BY728312.1 | - | 2 | 164 | |||
CN663576.1 | - | 2 | 84 |