Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176831.4 | NP_789801.1 | 7 | 1263 | ||
GenBank mRNA | AK036042.1 | - | 7 | 1246 | ||
AK039846.1 | - | 2 | 637 | |||
AK046751.1 | - | 4 | 1315 | |||
AK078825.1 | - | 7 | 1464 | |||
BC004779.1 | - | 7 | 1474 | |||
BC052928.1 | - | 6 | 1213 | 5255304 | ||
BC089351.1 | - | 6 | 1226 | 30603115 |