Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002358.3 | NP_002349.1 | 2 | 280 | ||
GenBank mRNA | AF394735.1 | - | 2 | 173 | ||
AJ000186.1 | - | 2 | 222 | |||
AK298228.1 | - | 2 | 255 | |||
AK313827.1 | - | 2 | 242 | |||
BC000356.2 | - | 2 | 230 | 2964388 | ||
BC005945.1 | - | 2 | 223 | 4132892 | ||
BC070283.1 | - | 2 | 192 | 6654207 |
Gene Symbol: MNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KU178192.1 | - | 2 | 243 |