Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173547.2 | NP_001167018.1 | 9 | 915 | ||
NM_001242615.2 | NP_001229544.1 | 8 | 862 | |||
NM_001242616.2 | NP_001229545.1 | 8 | 859 | |||
NM_001290738.1 | NP_001277667.1 | 7 | 781 | |||
NM_181516.6 | NP_852657.1 | 9 | 912 | |||
GenBank mRNA | AK004035.1 | - | 9 | 873 | ||
AK075615.1 | - | 9 | 869 | |||
AK160327.1 | - | 8 | 828 | |||
AK170662.1 | - | 8 | 1454 | |||
AK178858.1 | - | 6 | 331 | |||
AY231459.1 | - | 9 | 631 | |||
AY231460.1 | - | 8 | 578 | |||
AY258040.1 | - | 8 | 578 | |||
AY258041.1 | - | 9 | 631 | |||
BC015305.1 | - | 9 | 877 | 4221041 | ||
BC064023.1 | - | 1 | 207 | |||
CD807936.1 | - | 7 | 727 |