Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025790.2 | NP_080066.1 | 3 | 565 | ||
GenBank mRNA | AK002261.1 | - | 3 | 567 | ||
AK008624.1 | - | 3 | 523 | |||
AK144523.1 | - | 3 | 569 | |||
BC018165.1 | - | 3 | 546 | 3587125 | ||
BC096567.1 | - | 3 | 564 | 1314880 |