Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025794.2 | NP_080070.2 | 9, 10 | 1329 | ||
GenBank mRNA | AK002483.1 | - | 9, 10 | 1282 | ||
AK075673.1 | - | 9, 10 | 1280 | |||
AK141684.1 | - | 9, 10 | 1283 | |||
AK152624.1 | - | 9, 10 | 1298 | |||
AK169686.1 | - | 9, 10 | 1234 | |||
BC012522.1 | - | 9, 10 | 1278 | 2648103 | ||
BC057670.1 | - | 9, 10 | 1296 | 2810976 |