Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024180.5 | NP_077142.1 | 3, 4 | 489 | ||
GenBank mRNA | AK002583.1 | - | 3, 4 | 452 | ||
AK002824.1 | - | 2, 3 | 196 | |||
AK007548.1 | - | 2, 3 | 272 | |||
AK012681.1 | - | 2, 3 | 272 | |||
AK013455.1 | - | 3, 4 | 462 | |||
BC002146.1 | - | 3, 4 | 510 | 3484833 | ||
DV065903.1 | - | 3, 4 | 532 |