Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025796.3 | NP_080072.2 | 3 | 187 | ||
GenBank mRNA | AB049651.1 | - | 3 | 170 | ||
AK002421.1 | - | 3 | 167 | |||
AK008431.1 | - | 3 | 167 | |||
AK021173.1 | - | 3 | 167 | |||
AK041771.1 | - | 1 | 2465 | |||
AK165856.1 | - | 148 | ||||
AK167883.1 | - | 3 | 167 | |||
BC006029.1 | - | 1 | 2451 | |||
BC027018.1 | - | 153 | 5365357 | |||
BC037363.1 | - | 3 | 153 | 1514355 | ||
BC051471.1 | - | 3 | 164 | 1514101 | ||
BC055724.1 | - | 3 | 146 | 6411227 | ||
BC059722.1 | - | 3 | 166 | 6772871 | ||
BY112771.1 | - | 3 | 187 |