Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025799.4 | NP_080075.2 | 5 | 1184 | ||
GenBank mRNA | AK002675.1 | - | 5 | 1126 | ||
AK008653.1 | - | 5 | 1129 | |||
AK017443.1 | - | 5 | 1154 | |||
AK030320.1 | - | 5 | 1186 | |||
AK035607.1 | - | 5 | 1065 | |||
AK044790.1 | - | 5 | 1041 | |||
AK146295.1 | - | 5 | 1113 | |||
AK151204.1 | - | 5 | 1045 | |||
AK159288.1 | - | 5 | 1130 | |||
AK163042.1 | - | 3 | 577 | |||
AK218309.1 | - | 2 | 147 | |||
BC004039.1 | - | 5 | 1135 | 3496187 | ||
BC018181.1 | - | 5 | 1144 | 3983067 | ||
BC094449.1 | - | 5 | 1099 | 4222249 |