Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025799.4 | NP_080075.2 | 3 | 737 | ||
GenBank mRNA | AK002675.1 | - | 3 | 679 | ||
AK008653.1 | - | 3 | 682 | |||
AK030320.1 | - | 3 | 739 | |||
AK035607.1 | - | 3 | 618 | |||
AK044790.1 | - | 3 | 594 | |||
AK146295.1 | - | 3 | 666 | |||
AK151204.1 | - | 3 | 598 | |||
AK159288.1 | - | 3 | 683 | |||
AK204238.1 | - | 1 | 50 | |||
BC004039.1 | - | 3 | 688 | 3496187 | ||
BC094449.1 | - | 3 | 652 | 4222249 |