Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163689.1 | NP_001157161.1 | 2 | 497 | ||
NM_025802.3 | NP_080078.2 | 2 | 497 | |||
GenBank mRNA | AK002826.1 | - | 2 | 479 | ||
AK031609.1 | - | 2 | 455 | |||
AK076464.1 | - | 2 | 281 | |||
AK085926.1 | - | 2 | 481 | |||
AK150184.1 | - | 2 | 479 | |||
AY510273.1 | - | 2 | 478 | |||
AY731699.1 | - | 2 | 478 | |||
AY894805.1 | - | 2 | 398 | |||
BC019188.1 | - | 3 | 561 | 5027544 | ||
BC044737.1 | - | 3 | 543 | |||
BC044781.2 | - | 3 | 543 | 5151911 | ||
BC054825.1 | - | 3 | 538 | |||
BC064747.1 | - | 2 | 469 | 30024535 | ||
CF725975.1 | - | 2 | 487 |