Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037877.3 | NP_001032966.1 | 7 | 923 | ||
NM_001037878.3 | NP_001032967.1 | 7 | 923 | |||
NM_001286362.1 | NP_001273291.1 | 6 | 848 | |||
NM_001286363.1 | NP_001273292.1 | 6 | 848 | |||
NM_025804.3 | NP_080080.2 | 7 | 923 | |||
GenBank mRNA | AK003180.1 | - | 7 | 811 | ||
AK013754.1 | - | 7 | 612 | |||
AK045397.1 | - | 6 | 776 | |||
AK050274.1 | - | 7 | 851 | |||
AK075728.1 | - | 7 | 777 | |||
AK088360.1 | - | 7 | 811 | |||
AK089964.1 | - | 7 | 812 | |||
AK122430.1 | - | 7 | 778 | |||
AK148082.1 | - | 7 | 785 | |||
AK150232.1 | - | 7 | 810 | |||
AK150918.1 | - | 7 | 804 | |||
AK150942.1 | - | 7 | 797 | |||
AK151143.1 | - | 7 | 797 | |||
AK151558.1 | - | 7 | 804 | |||
AK152178.1 | - | 7 | 797 | |||
AK152494.1 | - | 7 | 797 | |||
AK161627.1 | - | 7 | 799 | |||
AK167837.1 | - | 7 | 797 | |||
AK168167.1 | - | 7 | 797 | |||
AK198411.1 | - | 4 | 234 | |||
BC016130.1 | - | 7 | 785 | |||
BC025071.1 | - | 7 | 784 | 3498003 | ||
BC046768.1 | - | 7 | 881 | 5707312 | ||
EU624323.1 | - | 6 | 686 | |||
U94988.1 | - | 7 | 784 |