Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025807.3 | NP_080083.2 | 5 | 1268 | ||
GenBank mRNA | AK004556.1 | - | 5 | 1269 | ||
AK004599.1 | - | 5 | 1447 | |||
AK004684.2 | - | 5 | 1326 | |||
AK015196.1 | - | 3 | 1045 | |||
BC034205.1 | - | 5 | 1223 | 4987055 | ||
BC055839.1 | - | 5 | 1226 | 4237811 |