Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290659.1 | NP_001277588.1 | 16 | 2996 | ||
NM_198294.3 | NP_938036.2 | 17 | 3172 | |||
GenBank mRNA | AB093299.1 | - | 6 | 1265 | ||
AK030022.1 | - | 14 | 2776 | |||
BC079914.1 | - | 16 | 2975 | 30668628 | ||
BC141367.1 | - | 16 | 2986 | 9055980 | ||
CX769146.1 | - | 1 | 72 |