Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024181.2 | NP_077143.2 | 3, 4 | 855 | ||
GenBank mRNA | AF255459.1 | - | 3, 4 | 532 | ||
AF314002.1 | - | 2, 3 | 368 | |||
AK004617.1 | - | 3, 4 | 610 | |||
AK013930.1 | - | 3, 4 | 551 | |||
AK016269.1 | - | 3, 4 | 855 | |||
AK028184.1 | - | 3, 4 | 595 | |||
AK128945.1 | - | 3, 4 | 610 | |||
AK165652.1 | - | 3, 4 | 613 | |||
AK166502.1 | - | 3, 4 | 565 | |||
AK169020.1 | - | 3, 4 | 622 | |||
AK179556.1 | - | 1, 2 | 104 | |||
BC033461.1 | - | 3, 4 | 594 | 4010341 |