Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025808.3 | NP_080084.2 | 6 | 620 | ||
GenBank mRNA | AK004561.2 | - | 6 | 620 | ||
AK005037.1 | - | 6 | 619 | |||
AK083411.1 | - | 6 | 624 | |||
AK132817.1 | - | 6 | 619 | |||
AK215859.1 | - | 2 | 180 | |||
BC034400.1 | - | 6 | 617 | 2651456 |