Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 2 | 427 | ||
NM_001145103.1 | NP_001138575.1 | 2 | 238 | |||
NM_005902.3 | NP_005893.1 | 2 | 662 | |||
XM_011521559.2 | XP_011519861.1 | 2 | 821 | |||
XM_011521560.1 | XP_011519862.1 | 2 | 601 | |||
GenBank mRNA | AB209616.1 | - | 2 | 247 | ||
AK290881.1 | - | 3 | 587 | |||
AK298139.1 | - | 2 | 238 | |||
AK316017.1 | - | 2 | 287 | |||
BC050743.1 | - | 2 | 580 | 6094328 | ||
DA453132.1 | - | 2 | 427 | |||
U68019.1 | - | 2 | 430 | |||
U76622.1 | - | 2 | 401 |