Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tril | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025817.4 | NP_080093.1 | 1 | 863 | ||
GenBank mRNA | AK004681.2 | - | 1 | 534 | ||
AK033401.1 | - | 1 | 526 | |||
AK051421.1 | - | 1 | 63 | |||
AK122338.1 | - | 1 | 451 | |||
BC043099.1 | - | 1 | 732 | 6406165 | ||
BC059224.1 | - | 1 | 732 | 6406165 |