Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1200014J11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025818.3 | NP_080094.3 | 9, 10 | 1032 | ||
GenBank mRNA | AK004766.1 | - | 9, 10 | 1020 | ||
AK011450.1 | - | 9, 10 | 1007 | |||
AK017496.1 | - | 9, 10 | 1027 | |||
AK032983.1 | - | 7, 8 | 739 | |||
AK033682.1 | - | 9, 10 | 1021 | |||
AK153152.1 | - | 9, 10 | 1020 | |||
AK153241.1 | - | 9, 10 | 1020 | |||
AK166563.1 | - | 7, 8 | 822 | |||
BC053720.1 | - | 7, 8 | 821 | |||
BC098495.1 | - | 9, 10 | 1004 | 30021633 | ||
BC118943.2 | - | 9, 10 | 1020 | 40092337 |